2-Fluoro-4-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51197G1 AS51197G5
ACTIAS51197G1EA 242 GBP
ACTIAS51197G1 ACTIAS51197G5
2-Fluoro-4-(tributylstannyl)pyridine
2-Fluoro-4-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
CAS Number: 457061-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR