2-Fluoro-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51188G1 AS51188G25 AS51188G5
ACTIAS51188G1EA 70 GBP
ACTIAS51188G1 ACTIAS51188G25 ACTIAS51188G5
2-Fluoro-3-(tributylstannyl)pyridine
2-Fluoro-3-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
CAS Number: 155533-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR