2-Fluoro-4-methoxyphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

AS22760G1 AS22760G5
ACTIAS22760G1EA 26 GBP
ACTIAS22760G1 ACTIAS22760G5
2-Fluoro-4-methoxyphenylboronic acid
2-Fluoro-4-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR