4-Quinazolone

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinazolin-4(3H)-one

ACTIAS20007G5EA 26 GBP
ACTIAS20007G5 ACTIAS20007G50 ACTIAS20007G250
4-Quinazolone
4-Quinazolone
Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR