5-Amino-1-methyl-1H-pyrazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

AS20973G1 AS20973G25 AS20973G5
ACTIAS20973G1EA 12 GBP
ACTIAS20973G1 ACTIAS20973G25 ACTIAS20973G5
5-Amino-1-methyl-1H-pyrazole
5-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00068156
CAS Number: 1192-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR