5-Bromo-2-chloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS50855G100 AS50855G25
ACTIAS50855G100EA 52 GBP
ACTIAS50855G100 ACTIAS50855G25
5-Bromo-2-chloropyridine
5-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 71 °C
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR