6-Methoxypicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methoxypyridine-2-carboxylic acid, 6-Methoxy-2-pyridinecarboxylic acid

AS22730G25 AS22730G5
ACTIAS22730G25EA 132 GBP
ACTIAS22730G25 ACTIAS22730G5
6-Methoxypicolinic acid
6-Methoxypicolinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD06800962
CAS Number: 26893-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR