Methyl picolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl pyridine-2-carboxylate

AS21647G100 AS21647G25
ACTIAS21647G100EA 84 GBP
ACTIAS21647G100 ACTIAS21647G25
Methyl picolinate
Methyl picolinate
Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00038038
CAS Number: 2459-07-6
EINECS: 219-545-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR