3-Chloropicolinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloropyridine-2-carboxylic acid

AS22714G1 AS22714G25 AS22714G5
ACTIAS22714G1EA 14 GBP
ACTIAS22714G1 ACTIAS22714G25 ACTIAS22714G5
3-Chloropicolinic acid
3-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD01646325
CAS Number: 57266-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR