2-Chloro-4-formylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde

AS33674G1 AS33674G5
ACTIAS33674G1EA 0 GBP
ACTIAS33674G1 ACTIAS33674G5
2-Chloro-4-formylpyridine
2-Chloro-4-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
MDL Number: MFCD06651557
CAS Number: 101066-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR