2-Amino-3,5-dibromo-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dibromo-6-methyl-2-pyridinamine

AS22533G100 AS22533G25
ACTIAS22533G100EA 0 GBP
ACTIAS22533G100 ACTIAS22533G25
2-Amino-3,5-dibromo-6-methylpyridine
2-Amino-3,5-dibromo-6-methylpyridine
Formula: C₆H₆Br₂N₂
MW: 265.94 g/mol
MDL Number: MFCD00068229
CAS Number: 91872-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR