2-Bromo-4-formylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-pyridinecarboxaldehyde, 2-Bromopyridine-4-carboxaldehyde

AS51242G1 AS51242G25 AS51242G5
ACTIAS51242G1EA 29 GBP
ACTIAS51242G1 ACTIAS51242G25 ACTIAS51242G5
2-Bromo-4-formylpyridine
2-Bromo-4-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
MDL Number: MFCD04039311
CAS Number: 118289-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR