DL-Tropic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid

AS23095G100 AS23095G25
ACTIAS23095G100EA 0 GBP
ACTIAS23095G100 ACTIAS23095G25
DL-Tropic acid
DL-Tropic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00004255
CAS Number: 529-64-6
EINECS: 208-465-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR