4-Chloro-2-iodobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS34292G1
AS34292G5
ACTIAS34292G1EA
0
GBP
ACTIAS34292G1
ACTIAS34292G5
4-Chloro-2-iodobenzoic acid
4-Chloro-2-iodobenzoic acid
Formula:
C₇H₄ClIO₂ MW: 282.46 g/mol |
MDL Number:
MFCD00060676 CAS Number: 13421-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...