Cyclopentanecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20668G100 AS20668G25
ACTIAS20668G100EA 0 GBP
ACTIAS20668G100 ACTIAS20668G25
Cyclopentanecarboxylic acid
Cyclopentanecarboxylic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001371
CAS Number: 3400-45-1
EINECS: 222-269-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR