3-Methyl-2-thenoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methylthiophene-2-carboxylic acid
AS20146G100
AS20146G25
ACTIAS20146G100EA
0
GBP
ACTIAS20146G100
ACTIAS20146G25
3-Methyl-2-thenoic acid
3-Methyl-2-thenoic acid
Formula:
C₆H₆O₂S MW: 142.18 g/mol |
MDL Number:
MFCD00005438 CAS Number: 23806-24-8 EINECS: 245-894-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...