Benzo[b]thiophene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Thianaphthene, 1-Benzothiophene
AS21333G100
AS21333G25
ACTIAS21333G100EA
0
GBP
ACTIAS21333G100
ACTIAS21333G25
Benzo[b]thiophene
Benzo[b]thiophene
Formula:
C₈H₆S MW: 134.2 g/mol |
MDL Number:
MFCD00005864 CAS Number: 95-15-8 EINECS: 202-395-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...