4-(4-Nitrophenyl)morpholine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21802G100
AS21802G25
ACTIAS21802G100EA
146
GBP
ACTIAS21802G100
ACTIAS21802G25
4-(4-Nitrophenyl)morpholine
4-(4-Nitrophenyl)morpholine
Formula:
C₁₀H₁₂N₂O₃ |
MDL Number:
MFCD00023317 CAS Number: 10389-51-2 EINECS: 233-851-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...