3-Ethylaniline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20424G100
AS20424G25
AS20424G5
ACTIAS20424G100EA
275
GBP
ACTIAS20424G100
ACTIAS20424G25
ACTIAS20424G5
3-Ethylaniline
3-Ethylaniline
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 212 °C (1013 hPa) Melting Pt: –8 °C Density: 0.97 g/cm³ (20 °C) Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007818 CAS Number: 587-02-0 EINECS: 209-594-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...