2-Iodo-5-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Iodo-2-methyl-4-nitrobenzene

AS24807G100 AS24807G25
ACTIAS24807G100EA 167 GBP
ACTIAS24807G100 ACTIAS24807G25
2-Iodo-5-nitrotoluene
2-Iodo-5-nitrotoluene
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051517
CAS Number: 5326-38-5
EINECS: 226-204-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR