4-Bromo-3-methylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS34528G1 AS34528G5
ACTIAS34528G1EA 17 GBP
ACTIAS34528G1 ACTIAS34528G5
4-Bromo-3-methylbenzaldehyde
4-Bromo-3-methylbenzaldehyde
Formula: C₈H₇BrO
MDL Number: MFCD04971197
CAS Number: 78775-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR