5-Bromo-2-methylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32510G1 AS32510G5
ACTIAS32510G1EA 0 GBP
ACTIAS32510G1 ACTIAS32510G5
5-Bromo-2-methylbenzaldehyde
5-Bromo-2-methylbenzaldehyde
Formula: C₈H₇BrO
MDL Number: MFCD05662390
CAS Number: 90050-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR