6-Chlorooxindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Chloro-2-oxindole

AS20391G100 AS20391G25
ACTIAS20391G100EA 137 GBP
ACTIAS20391G100 ACTIAS20391G25
6-Chlorooxindole
6-Chlorooxindole
Formula: C₈H₆ClNO
MW: 167.59 g/mol
MDL Number: MFCD00209962
CAS Number: 56341-37-8
EINECS: 421-330-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR