2-Fluoro-4-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22674G100 AS22674G25
ACTIAS22674G100EA 165 GBP
ACTIAS22674G100 ACTIAS22674G25
2-Fluoro-4-nitrobenzoic acid
2-Fluoro-4-nitrobenzoic acid
Formula: C₇H₄FNO₄
MDL Number: MFCD00275565
CAS Number: 403-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR