3-Fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

ACTIAS20229G100EA 0 GBP
ACTIAS20229G100 ACTIAS20229G500
3-Fluorobenzoic acid
3-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR