2,4,6-Trifluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22442G100 AS22442G25
ACTIAS22442G100EA 211 GBP
ACTIAS22442G100 ACTIAS22442G25
2,4,6-Trifluorobenzonitrile
2,4,6-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: 57…58 °C
MDL Number: MFCD00042399
CAS Number: 96606-37-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR