3-Chloro-2-fluorobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24707G100
AS24707G25
AS24707G5
ACTIAS24707G100EA
191
GBP
ACTIAS24707G100
ACTIAS24707G25
ACTIAS24707G5
3-Chloro-2-fluorobenzonitrile
3-Chloro-2-fluorobenzonitrile
Formula:
C₇H₃ClFN MW: 155.56 g/mol |
MDL Number:
MFCD00042206 CAS Number: 94087-40-8 EINECS: 301-934-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...