4-Chloro-2-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS24336G100 AS24336G25
ACTIAS24336G100EA 109 GBP
ACTIAS24336G100 ACTIAS24336G25
4-Chloro-2-fluorobenzaldehyde
4-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD00143282
CAS Number: 61072-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR