3-Chloro-4-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20147G100 AS20147G25
ACTIAS20147G100EA 100 GBP
ACTIAS20147G100 ACTIAS20147G25
3-Chloro-4-fluorobenzaldehyde
3-Chloro-4-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 66 °C (2 mmHg)
Melting Pt: 44…46 °C
MDL Number: MFCD00011735
CAS Number: 34328-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR