2-Chloro-5-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23163G100 AS23163G25
ACTIAS23163G100EA 78 GBP
ACTIAS23163G100 ACTIAS23163G25
2-Chloro-5-nitrobenzonitrile
2-Chloro-5-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Boiling Pt: 119…122 °C (0.6 mmHg)
Melting Pt: 108 °C
MDL Number: MFCD00007292
CAS Number: 16588-02-6
EINECS: 240-643-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR