2,5-Dichlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24977G100 AS24977G25
ACTIAS24977G100EA 0 GBP
ACTIAS24977G100 ACTIAS24977G25
2,5-Dichlorobenzonitrile
2,5-Dichlorobenzonitrile
Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 130…132 °C
MDL Number: MFCD00019741
CAS Number: 21663-61-6
EINECS: 244-518-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR