4-Nitro-m-anisic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methoxy-4-nitrobenzoic acid
AS22980G100
AS22980G25
ACTIAS22980G100EA
105
GBP
ACTIAS22980G100
ACTIAS22980G25
4-Nitro-m-anisic acid
4-Nitro-m-anisic acid
Formula:
C₈H₇NO₅ MW: 197.15 g/mol Boiling Pt: 405 °C (1013 hPa) Melting Pt: 233 °C |
MDL Number:
MFCD00007353 CAS Number: 5081-36-7 EINECS: 225-793-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...