1-Bromo-3-(trifluoromethoxy)benzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trifluoromethoxy)bromobenzene

AS20315G100 AS20315G25
ACTIAS20315G100EA 44 GBP
ACTIAS20315G100 ACTIAS20315G25
1-Bromo-3-(trifluoromethoxy)benzene
1-Bromo-3-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 78…81 °C (55 mmHg)
Density: 1.635 g/cm³ (20 °C)
MDL Number: MFCD00040943
CAS Number: 2252-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR