4-(Trifluoromethyl)phenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride

AS21363G100 AS21363G25
ACTIAS21363G100EA 0 GBP
ACTIAS21363G100 ACTIAS21363G25
4-(Trifluoromethyl)phenylhydrazinium chloride
4-(Trifluoromethyl)phenylhydrazinium chloride
Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
MDL Number: MFCD00204233
CAS Number: 2923-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR