m-Tolylhydrazine hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21207G100 AS21207G25
ACTIAS21207G100EA 0 GBP
ACTIAS21207G100 ACTIAS21207G25
m-Tolylhydrazine hydrochloride
m-Tolylhydrazinium chloride
Formula: C₇H₁₀N₂·HCl
MW: 158.63 g/mol
MDL Number: MFCD00012932
CAS Number: 637-04-7
EINECS: 211-276-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR