5-Bromo-2-chlorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20092G100 AS20092G25
ACTIAS20092G100EA 49 GBP
ACTIAS20092G100 ACTIAS20092G25
5-Bromo-2-chlorobenzoic acid
5-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR