4-Cyclohexylphenol

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxyphenylcyclohexane

ACTIAS24162G100EA 138 GBP
ACTIAS24162G100
4-Cyclohexylphenol
4-Cyclohexylphenol
Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD00035699
CAS Number: 1131-60-8
EINECS: 214-465-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR