3-Amino-4-iodopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine

AS13019G1 AS13019G5
ACTIAS13019G1EA 0 GBP
ACTIAS13019G1 ACTIAS13019G5
3-Amino-4-iodopyridine
3-Amino-4-iodopyridine
Formula: C₅H₅IN₂
MW: 220.01 g/mol
MDL Number: MFCD04971334
CAS Number: 105752-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR