4-Iodo-1-methylimidazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodo-1-methyl-1H-imidazole

AS12911G1 AS12911G5
ACTIAS12911G1EA 24 GBP
ACTIAS12911G1 ACTIAS12911G5
4-Iodo-1-methylimidazole
4-Iodo-1-methylimidazole
Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD01632214
CAS Number: 71759-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR