3-Nitro-5-(trifluoromethyl)phenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Hydroxy-5-nitrobenzotrifluoride

AS20238G25 AS20238G5
ACTIAS20238G25EA 303 GBP
ACTIAS20238G25 ACTIAS20238G5
3-Nitro-5-(trifluoromethyl)phenol
3-Nitro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
MDL Number: MFCD04973779
CAS Number: 349-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR