2-Fluoro-5-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Fluoro-2-methoxy-4-nitrobenzene, 4-Fluoro-3-methoxynitrobenzene

AS31499G1 AS31499G5
ACTIAS31499G1EA 0 GBP
ACTIAS31499G1 ACTIAS31499G5
2-Fluoro-5-nitroanisole
2-Fluoro-5-nitroanisole
Formula: C₇H₆FNO₃
MW: 171.13 g/mol
MDL Number: MFCD07782081
CAS Number: 454-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR