4-Fluorophenyl acetate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluorophenyl Acetate, 1-Acetoxy-4-fluorobenzene, Acetic acid 4-Fluorophenyl acetate, 1-Acetoxy-4-fluorobenzene

AS24661G100 AS24661G25
ACTIAS24661G100EA 98 GBP
ACTIAS24661G100 ACTIAS24661G25
4-Fluorophenyl acetate
4-Fluorophenyl acetate
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 58.5…59.5 °C (1 mmHg)
Density: 1,17 g/cm³ (25 °C)
MDL Number: MFCD00041361
CAS Number: 405-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR