Methyl 4-fluoro-3-nitrobenzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS33777G100
AS33777G5
ACTIAS33777G100EA
83
GBP
ACTIAS33777G100
ACTIAS33777G5
Methyl 4-fluoro-3-nitrobenzoate
Methyl 4-fluoro-3-nitrobenzoate
Formula:
C₈H₆FNO₄ |
MDL Number:
MFCD00196161 CAS Number: 329-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...