2-Fluoronicotinonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Cyano-2-fluoropyridine

AS24819G100 AS24819G25 AS24819G5
ACTIAS24819G100EA 0 GBP
ACTIAS24819G100 ACTIAS24819G25 ACTIAS24819G5
2-Fluoronicotinonitrile
2-Fluoronicotinonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 30…30.5 °C
MDL Number: MFCD03095082
CAS Number: 3939-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR