3-Cyano-4-methylpyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
AS21374G25
AS21374G5
ACTIAS21374G25EA
0
GBP
ACTIAS21374G25
ACTIAS21374G5
3-Cyano-4-methylpyridine
3-Cyano-4-methylpyridine
Formula:
C₇H₆N₂ MW: 118.14 g/mol |
MDL Number:
MFCD00234272 CAS Number: 5444-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...