2-Amino-5-chloro-3-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Chloro-3-fluoro-2-pyridylamine, 5-Chloro-3-fluoropyridin-2-amine

AS20446G1 AS20446G25 AS20446G5
ACTIAS20446G1EA 14 GBP
ACTIAS20446G1 ACTIAS20446G25 ACTIAS20446G5
2-Amino-5-chloro-3-fluoropyridine
2-Amino-5-chloro-3-fluoropyridine
Formula: C₅H₄ClFN₂
MW: 146.55 g/mol
MDL Number: MFCD09878434
CAS Number: 246847-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR