2'-Chloro-5'-fluoroacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20658G25 AS20658G5
ACTIAS20658G25EA 227 GBP
ACTIAS20658G25 ACTIAS20658G5
2'-Chloro-5'-fluoroacetophenone
2'-Chloro-5'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD00042574
CAS Number: 2965-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR