2-Chlorobenzamidine hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chlorobenzene-1-carboximidamide hydrochloride, 2-Chloro-benzamidine hydrochloride, 2-Chlorophenylamidine hydrochloride

AS21117G1 AS21117G5
ACTIAS21117G1EA 79 GBP
ACTIAS21117G1 ACTIAS21117G5
2-Chlorobenzamidine hydrochloride
2-Chlorobenzamidine hydrochloride
Formula: C₇H₈Cl₂N₂
MW: 191.06 g/mol
MDL Number: MFCD00275607
CAS Number: 18637-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR