4-Acetylbenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl p-cyanophenyl ketone, 4'-Cyanoacetophenone, 4-Acetylbenzenecarbonitrile

AS23893G100 AS23893G25
ACTIAS23893G100EA 0 GBP
ACTIAS23893G100 ACTIAS23893G25
4-Acetylbenzonitrile
4-Acetylbenzonitrile
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 110…112 °C (1 mmHg)
Melting Pt: 57…58 °C
MDL Number: MFCD00001825
CAS Number: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR