3-Acetylbenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3'-Cyanoacetophenone, 3-Acetylbenzenecarbonitrile, Methyl m-cyanophenyl ketone
AS20564G25
AS20564G5
ACTIAS20564G25EA
0
GBP
ACTIAS20564G25
ACTIAS20564G5
3-Acetylbenzonitrile
3-Acetylbenzonitrile
Formula:
C₉H₇NO MW: 145.16 g/mol Boiling Pt: 120 °C (5 mmHg) Melting Pt: 98…100 °C |
MDL Number:
MFCD00001806 CAS Number: 6136-68-1 EINECS: 228-110-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...